missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ I-BET151, 95%

Catalog No. AC467841000
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC467841000 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC467841000 Supplier Thermo Scientific™ Supplier No. 467841000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material I-BET151
CAS 1300031-49-5
Synonym 7-(3,5-Dimethyl-4-isoxazolyl)-1,3-dihydro-8-methoxy-1-[(1 R)-1-(2-pyridinyl)ethyl]-2 H-imidazo[4,5-c]quinolin-2-one
Quantity 100 mg
Molecular Formula C23 H21 N5 O3
Formula Weight 415.45
Physical Form Solid
Product Type Reagent
Purity 95%