missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyrTrp Cell Dissociation Reagent
Recombinant and animal-derived component-free alternative to traditional porcine trypsin
$88.75 - $176.67
Specifications
| Content And Storage | -20 °C |
|---|---|
| Product Type | Cell Culture Dissociation Reagent |
| For Use With | Attachment-dependent cell types MRC-5, Vero, CHO K-1 cell lines grown with or without serum |
Description
- Animal-derived component-free (ADCF)
- Dye-free
- Suitable for a variety of applications and cell types
- Effective cell dissociation
- Does not impact cell line viability and doubling times
- For cell-based vaccine production and stem cell/primary cell/gene therapy cell culture processes
- For use with attachment-dependent cell types MRC-5, Vero, CHO K-1 cell lines grown with or without serum
Specifications
| -20 °C | |
| Cell Culture Dissociation Reagent | |
| Attachment-dependent cell types MRC-5, Vero, CHO K-1 cell lines grown with or without serum |
Safety and Handling
ShelfLife : 12 months from manufacturing date when stored at -20°C