missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyrTrp Cell Dissociation Reagent

Catalog No. SV3020902
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SV3020902 500 mL
SV3020901 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SV3020902 Supplier Cytiva Supplier No. SV30209.02
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Recombinant and animal-derived component-free alternative to traditional porcine trypsin

  • Animal-derived component-free (ADCF)
  • Dye-free
  • Suitable for a variety of applications and cell types
  • Effective cell dissociation
  • Does not impact cell line viability and doubling times
  • For cell-based vaccine production and stem cell/primary cell/gene therapy cell culture processes
  • For use with attachment-dependent cell types MRC-5, Vero, CHO K-1 cell lines grown with or without serum

Specifications

Content And Storage -20 °C
Product Type Cell Culture Dissociation Reagent
For Use With Attachment-dependent cell types MRC-5, Vero, CHO K-1 cell lines grown with or without serum
Quantity 500 mL