Learn More
Thermo Scientific™ Hypersil™ MOS-2 C8 HPLC Columns
Description
For reliable, affordable analysis of more hydrophobic analytes such as small peptides and proteins, pharmaceuticals, steroids, and environmental samples, use Thermo Scientific™ Hypersil™ MOS-2 C8 HPLC Columns.
Hypersil stationary phases are recognized as an HPLC industry standard. A proven effective analytical tool, classical Hypersil phases are well established and referenced in HPLC methods. Quality is assured by manufacture under ISO 9001-2000 guidelines.
For separations requiring improved chromatographic performance, use Thermo Scientific™ Hypersil GOLD™ C8 columns as an alternative.
- Reliable, less retentive phase
- Long column lifetimes, even under basic conditions
- Manufactured under ISO 9001-2000 quality guidelines
- Affordable
Specifications
Specifications
| Column Format | Analytical Column |
| Diameter (Metric) | 4.6 mm |
| Length (Metric) | 150 mm |
| Max. Pressure | 5800 psi (400 bar) |
| Particle Size | 5 μm |
| pH | 2 to 8 |
| Pore Size | 120 Å |
| Surface Area | 170 m²/g |
| Temperature | 60°C |
| USP Type | L7 |
| Show More |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.