Learn More
Thermo Scientific™ Hypersil™ Green PAH HPLC Columns
Use for rapid analysis of polyaromatic hydrocarbons (PAHs). They feature tailored alkyl-bonded silica with high carbon loading for excellent performance.
$1294.85 - $2877.42
Specifications
Max. Pressure 5800 psi (400 bar)
Pore Size 120 Å
Surface Area 170 m²/g
Temperature 60°C
Endcapped Yes
View More Specs
Products 12
Catalog Number Mfr. No. Length (Metric) Diameter (Metric) Particle Size Stationary Phase Price Quantity  
Catalog Number Mfr. No. Length (Metric) Diameter (Metric) Particle Size Stationary Phase Price Quantity  
31103102130
View Documents
Thermo Scientific™
31103102130
100 mm 2.1 mm 3 μm Phenyl (Ph)
Each for $2,679.87
Only null left
31103103030
View Documents
Thermo Scientific™
31103103030
100 mm 3 mm 3 μm Phenyl (Ph)
Each for $2,877.42
Only null left
31103104630
View Documents
Thermo Scientific™
31103104630
100 mm 4.6 mm 3 μm Phenyl (Ph)
Each for $2,551.79
Only null left
31103152130
View Documents
Thermo Scientific™
31103152130
150 mm 2.1 mm 3 μm Phenyl (Ph)
Each for $2,834.06
Only null left
31103154630
View Documents
Thermo Scientific™
31103154630
150 mm 4.6 mm 3 μm Phenyl (Ph)
Each for $2,688.57
Only null left
31105254630
View Documents
Thermo Scientific™
31105254630
250 mm 4.6 mm 5 μm Phenyl (Ph)
Each for $2,566.42
Only null left
31105154630
View Documents
Thermo Scientific™
31105154630
150 mm 4.6 mm 5 μm Phenyl (Ph)
Each for $2,450.00
Only null left
31105104630
View Documents
Thermo Scientific™
31105104630
100 mm 4.6 mm 5 μm Proprietary
Each for $2,108.96
Only null left
31105014001
View Documents
Thermo Scientific™
31105014001
10 mm 4 mm 5 μm Proprietary
Pack of 4 Each for $1,294.85
Only null left
31103014001
View Documents
Thermo Scientific™
31103014001
10 mm 4 mm 3 μm Phenyl (Ph)
Pack of 4 Each for $1,428.68
Only null left
31103013001
View Documents
Thermo Scientific™
31103013001
10 mm 3 mm 3 μm Proprietary
Pack of 4 Each for $1,428.68
Only null left
31103012101
View Documents
Thermo Scientific™
31103012101
10 mm 2.1 mm 3 μm Phenyl (Ph)
Pack of 4 Each for $1,428.68
Only null left
Description
Description
Rapidly analyze polyaromatic hydrocarbons (PAHs) with Thermo Scientific™ Hypersil™ Green Green PAH LC Columns. Featuring specially tailored alkyl-bonded silica with high carbon loading, these short, fast columns can analyze 16 PAHs in four minutes. Optimized for EPA Method 610: Polynuclear Aromatic Hydrocarbons, these columns provide excellent performance.
Wide Variety of Column Dimensions to Meet Your PAH Analysis Needs
  • 3.0μm particle size available in 2.1, 3.0 and 4.6mm diameters and 10, 100 and 150mm lengths
  • 5.0μm particle size available in 4.6mm diameter and 10, 100, 150 and 250mm lengths
Specifications
Specifications
5800 psi (400 bar)
170 m²/g
Yes
120 Å
60°C
Reversed Phase
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.