Learn More
Thermo Scientific™ Hypersil GOLD™ Peptide HPLC Column
Hypersil GOLD™ Peptide performance is assured by our dual quality confirmation, which is performed on both neutrals compounds as well as on a Cytochrome C digest.
Supplier: Thermo Scientific™ 26002102130
Description
Resolve peptide challenges with one column - NEW Hypersil GOLD Peptide.
Achieve consistent retention of all peptides, including hydrophilic ones, and resolve deamidated species. Designed for stability and lot-to-lot consistency, this column meets the needs of biopharmaceutical product development and manufacturing control.
Hypersil GOLD Peptide column performance is assured through dual quality confirmation, including a peptide-based QC test, ensures confidence in ultrapure silica, ligand bonding, and column packing for unparalleled sequence coverage and higher peak capacity compared to other peptide columns.
Consistent, Reliable Results
Enjoy excellent reproducibility with minimal RSD%, ensuring consistency from column-to-column and lot-to-lot.
Optimized Peptide Retention - Retain More, Discover More
Our column uniquely holds onto hydrophilic peptides, ensuring no critical data is missed.
Exceptional Peak Capacity - Separate with Superiority
Achieve higher peak capacity for detailed and clear separations, ensuring every component is distinctly resolved.
Advanced PTM Analysis - Decipher Complex Modifications
Efficiently separate key post-translational modifications like deamidation, crucial for in-depth protein analysis. Excellent peak shape for peptides using either FA- or TFA-containing mobile phases.
Unparalleled Sequence Coverage - See the Full Picture
High sequence coverage provides a comprehensive analysis, revealing the true complexity of your samples.
Specifications
| 2.1 mm | |
| 100 mm | |
| 1.9 μm | |
| 175 Å | |
| 220 m²/g | |
| L1 | |
| Spherical, Fully Porous, Ultrapure Silica | |
| Reversed Phase | |
| Reversed Phase | |
| Analytical Column | |
| Peptide |
| LC/MS | |
| 21756 psi (1500 bar) | |
| 2 to 11 | |
| 1 Each | |
| 60°C | |
| Hypersil GOLD | |
| 11% | |
| Yes | |
| Silica | |
| Reversed Phase HPLC |