missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Hypersil GOLD™ C4 HPLC Columns

Catalog No. p-4523052
Change view
Click to view available options
Length (Metric):
10 mm
50 mm
100 mm
150 mm
250 mm
Diameter (Metric):
2.1 mm
3 mm
4.6 mm
Particle Size:
1.9 μm
3 μm
5 μm
Catalog No. Length (Metric) Diameter (Metric) Particle Size
25505154630 150 mm 4.6 mm 5 μm
25505254630 250 mm 4.6 mm 5 μm
25502052130 50 mm 2.1 mm 1.9 μm
25502102130 100 mm 2.1 mm 1.9 μm
25502152130 150 mm 2.1 mm 1.9 μm
25503012101 10 mm 2.1 mm 3 μm
25503013001 10 mm 3 mm 3 μm
25503052130 50 mm 2.1 mm 3 μm
25503154630 150 mm 4.6 mm 3 μm
9 options

Catalog No. 25505154630

Supplier: Thermo Scientific™ 25505154630

Only null left

Featuring a lower hydrophobicity than C18 or C8 columns, Thermo Scientific™ Hypersil GOLD™ C4 columns provide outstanding sensitivity and high efficiency.

For very hydrophobic analytes, achieve faster separations with excellent peak shapes using Thermo Scientific™ Hypersil GOLD™ C4 HPLC columns. Featuring a lower hydrophobicity than C18 or C8 columns, Hypersil GOLD C4 columns provide outstanding sensitivity and high efficiency.

Similar selectivity, less retention than C18 or C8 columns
  • Successful retention and separation of very hydrophobic analytes with this short alkyl chain length, low hydrophobicity column
  • Ideal for analysis of hydrophobic polypeptides and proteins
Superior Performance
  • Highly symmetrical peak shapes
  • 1.9μm particle size columns improve speed and efficiency
About the Hypersil GOLD family of HPLC columns

Meet the challenges of modern chromatography with state-of-the-art columns that provide outstanding performance and consistent separations. Twelve different chemistries are available so you can optimize separations and maximize productivity. Get outstanding peak shapes in reversed phase, ion exchange, HILIC, or normal phase chromatography.

  • Excellent peak symmetry
  • Narrow peaks for outstanding efficiency
  • Enhanced sensitivity and improved resolution
  • Exceptional reproducibility for reliable chromatography, column after column
  • Solutions for high-throughput screening
  • 1.9, 3, and 5μm particle sizes for all your separation needs

Specifications

Column Format Analytical Column
Diameter (Metric) 4.6 mm
Length (Metric) 150 mm
Max. Pressure 5800 psi (400 bar)
Particle Size 5 μm
pH 2 to 8
Pore Size 175 Å
Surface Area 220 m²/g
Temperature 60°C
USP Type L26
Stationary Phase C4
Product Line Hypersil GOLD
Packing Material Spherical, Fully Porous, Ultrapure Silica
Carbon Load 5%
Column Type Reversed Phase
Endcapped Yes
Phase Reversed Phase
Show More Show Less

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.