missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ HyPerforma Tank Liners, CX3-9 Liner

Tank liner system with top train is composed of CX3-9 single web, multi-layer plastic film, with dimensions optimized for drums and commonly used industry standard cylindrical tanks. They feature an open top for maximum recovery, with drum dolly available to provide mobility.
$182.63 - $588.94
Specifications
| Outer Packaging | Cardboard Cartons |
|---|---|
| Protective Inner Packaging | Packed and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area |
| Sterilization Method | Irradiated |
| Product Line | HyPerforma |
| Film Description | CX3-9 |
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
SH3064710
![]() |
Thermo Scientific™
SH30647.10 |
378 L |
Each for $182.63
|
|
|||||
SH3064711
![]() |
Thermo Scientific™
SH30647.11 |
757 L |
Each for $588.94
|
|
|||||
SH3064718
![]() |
Thermo Scientific™
SH30647.18 |
19 L |
Each for $198.00
|
|
|||||
SH3064719
![]() |
Thermo Scientific™
SH30647.19 |
28 L |
Each for $214.81
|
|
|||||
SH3064720
![]() |
Thermo Scientific™
SH30647.20 |
38 L |
Each for $187.78
|
|
|||||
Description
- Minimizes bioburden due to construction in an ISO 7 cleanroom under sterile cGMP conditions.
- Eliminates post-use cleaning steps required with re-useable containers and reduces cross-contamination risks.
Applications:
- Removes the need for tank cleaning and helps reduce cycle times.
- Facilitates hydration of powdered media and buffers, and pooling of non-sterile solutions and fluids.
Specifications
| Cardboard Cartons | |
| Irradiated | |
| CX3-9 |
| Packed and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area | |
| HyPerforma |