Learn More
Thermo Scientific™ HyPerforma Tank Liners, CX3-9 Liner
GREENER_CHOICE
Catalog No. SH3064720
Change view
Click to view available options
Capacity (Metric):
19 L
28 L
38 L
378 L
757 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
SH3064720 38 L
SH3064710 378 L
SH3064711 757 L
SH3064718 19 L
SH3064719 28 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. SH3064720 Supplier Thermo Scientific™ Supplier No. SH30647.20
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Tank liner system with top train is composed of CX3-9 single web, multi-layer plastic film, with dimensions optimized for drums and commonly used industry standard cylindrical tanks. They feature an open top for maximum recovery, with drum dolly available to provide mobility.
  • Minimizes bioburden due to construction in an ISO 7 cleanroom under sterile cGMP conditions.
  • Eliminates post-use cleaning steps required with re-useable containers and reduces cross-contamination risks.
Applications:
  • Removes the need for tank cleaning and helps reduce cycle times.
  • Facilitates hydration of powdered media and buffers, and pooling of non-sterile solutions and fluids.
Specifications
Capacity (Metric) 38 L
Capacity (English) 10 gal.
Outer Packaging Cardboard Cartons
Protective Inner Packaging Packed and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area
Sterilization Method Irradiated
Product Line HyPerforma
Film Description CX3-9