missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hygromycin B, 92.30%, For HPLC analysis, MP Biomedicals™
$78.96 - $119.42
Chemical Identifiers
| CAS | 31282-04-9 |
|---|---|
| Molecular Formula | C20H37N3O13 |
| Molecular Weight (g/mol) | 527.524 |
| InChI Key | GRRNUXAQVGOGFE-BBMONYMYSA-N |
| Synonym | Hygromix, Antihelmycin |
| PubChem CID | 134129613 |
| IUPAC Name | (3'S,3aS,4S,4'S,5'R,6R,6'R,7R,7aS)-4-[(1R,2S,3R,5S,6S)-3-amino-2,6-dihydroxy-5-(methylamino)cyclohexyl]oxy-6'-[(1S)-1-amino-2-hydroxyethyl]-6-(hydroxymethyl)spiro[4,6,7,7a-tetrahydro-3aH-[1,3]dioxolo[4,5-c]pyran-2,2'-oxane]-3',4',5',7-tetrol |
| SMILES | CNC1CC(C(C(C1O)OC2C3C(C(C(O2)CO)O)OC4(O3)C(C(C(C(O4)C(CO)N)O)O)O)O)N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
ICN19417050
|
N/A | 50 mg |
Each for $78.96
|
|
|||||
|
ICN19417080
|
N/A | 100 mg |
Each for $119.42
|
|
|||||
Chemical Identifiers
| 31282-04-9 | |
| 527.524 | |
| Hygromix, Antihelmycin | |
| (3'S,3aS,4S,4'S,5'R,6R,6'R,7R,7aS)-4-[(1R,2S,3R,5S,6S)-3-amino-2,6-dihydroxy-5-(methylamino)cyclohexyl]oxy-6'-[(1S)-1-amino-2-hydroxyethyl]-6-(hydroxymethyl)spiro[4,6,7,7a-tetrahydro-3aH-[1,3]dioxolo[4,5-c]pyran-2,2'-oxane]-3',4',5',7-tetrol |
| C20H37N3O13 | |
| GRRNUXAQVGOGFE-BBMONYMYSA-N | |
| 134129613 | |
| CNC1CC(C(C(C1O)OC2C3C(C(C(O2)CO)O)OC4(O3)C(C(C(C(O4)C(CO)N)O)O)O)O)N |
Specifications
| Hygromycin B | |
| 31282-04-9 | |
| C20H37N3O13 | |
| GRRNUXAQVGOGFE-BBMONYMYSA-N | |
| (3'S,3aS,4S,4'S,5'R,6R,6'R,7R,7aS)-4-[(1R,2S,3R,5S,6S)-3-amino-2,6-dihydroxy-5-(methylamino)cyclohexyl]oxy-6'-[(1S)-1-amino-2-hydroxyethyl]-6-(hydroxymethyl)spiro[4,6,7,7a-tetrahydro-3aH-[1,3]dioxolo[4,5-c]pyran-2,2'-oxane]-3',4',5',7-tetrol | |
| 134129613 | |
| 92.3% |
| Liquid | |
| Brown to Red | |
| Hygromix, Antihelmycin | |
| CNC1CC(C(C(C1O)OC2C3C(C(C(O2)CO)O)OC4(O3)C(C(C(C(O4)C(CO)N)O)O)O)O)N | |
| 527.524 | |
| 527.5 | |
| Hygromycin B, For HPLC analysis |