missing translation for 'onlineSavingsMsg'
Learn More

Hygromycin B, 92.30%, For HPLC analysis, MP Biomedicals™

Catalog No. ICN19417050
Change view
Click to view available options
Quantity:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19417050 50 mg
ICN19417080 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19417050 Supplier MP Biomedicals Inc Supplier No. 0219417050

Chemical Identifiers

CAS 31282-04-9
Molecular Formula C20H37N3O13
Molecular Weight (g/mol) 527.524
InChI Key GRRNUXAQVGOGFE-BBMONYMYSA-N
Synonym Hygromix, Antihelmycin
PubChem CID 134129613
IUPAC Name (3'S,3aS,4S,4'S,5'R,6R,6'R,7R,7aS)-4-[(1R,2S,3R,5S,6S)-3-amino-2,6-dihydroxy-5-(methylamino)cyclohexyl]oxy-6'-[(1S)-1-amino-2-hydroxyethyl]-6-(hydroxymethyl)spiro[4,6,7,7a-tetrahydro-3aH-[1,3]dioxolo[4,5-c]pyran-2,2'-oxane]-3',4',5',7-tetrol
SMILES CNC1CC(C(C(C1O)OC2C3C(C(C(O2)CO)O)OC4(O3)C(C(C(C(O4)C(CO)N)O)O)O)O)N

Specifications

Type Hygromycin B
Physical Form Liquid
Color Brown to Red
Quantity 50 mg
Formula Weight 527.5
Percent Purity 92.3%
Chemical Name or Material Hygromycin B, For HPLC analysis