missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Hydroxyurea | |
| 140°C | |
| CH4N2O2 | |
| 25 g | |
| hydroxycarbamide, n-hydroxyurea, hydrea, 1-hydroxyurea, oxyurea, carbamoyl oxime, biosupressin, hydroxycarbamine, onco-carbide, carbamohydroxamic acid | |
| C(=O)(N)NO | |
| 76.055 | |
| CHEBI:44423 | |
| ≥97.0% (N) |
| 127-07-1 | |
| White | |
| MFCD00007943 | |
| 2811 | |
| VSNHCAURESNICA-UHFFFAOYSA-N | |
| hydroxyurea | |
| 3657 | |
| 76.06 | |
| Crystalline Powder |
Chemical Identifiers
| 127-07-1 | |
| 76.055 | |
| VSNHCAURESNICA-UHFFFAOYSA-N | |
| 3657 | |
| hydroxyurea |
| CH4N2O2 | |
| MFCD00007943 | |
| hydroxycarbamide, n-hydroxyurea, hydrea, 1-hydroxyurea, oxyurea, carbamoyl oxime, biosupressin, hydroxycarbamine, onco-carbide, carbamohydroxamic acid | |
| CHEBI:44423 | |
| C(=O)(N)NO |
Safety and Handling
RTECSNumber : YT4900000
TSCA : No