missing translation for 'onlineSavingsMsg'
Learn More

Hydroxyurea 97.0+%, TCI America™

Catalog No. p-7130480
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H03105G 5 g
H031025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H03105G Supplier TCI America Supplier No. H03105G
Only null left

Chemical Identifiers

CAS 127-07-1
Molecular Formula CH4N2O2
Molecular Weight (g/mol) 76.055
MDL Number MFCD00007943
InChI Key VSNHCAURESNICA-UHFFFAOYSA-N
Synonym hydroxycarbamide, n-hydroxyurea, hydrea, 1-hydroxyurea, oxyurea, carbamoyl oxime, biosupressin, hydroxycarbamine, onco-carbide, carbamohydroxamic acid
PubChem CID 3657
ChEBI CHEBI:44423
IUPAC Name hydroxyurea
SMILES C(=O)(N)NO

Specifications

Melting Point 140°C
Color White
Quantity 5 g
UN Number 2811
Formula Weight 76.06
Percent Purity ≥97.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Hydroxyurea
RTECSNumber YT4900000
TSCA No