Learn More
Hydroxylamine Hydrochloride, For Mercury, Certified, 50% (w/v), LabChem™
Supplier: LabChem LC155501
Specifications
| Hydroxylamine Hydrochloride | |
| For Mercury | |
| 50,50 | |
| H3NO·HCl | |
| UN3287 | |
| Soluble in water | |
| Cl.NO | |
| 69.49 | |
| CHEBI:5807 | |
| 50% (w/v) | |
| Passes Test | |
| Gaseous Ammonia; Nitrogen oxides; Hydrogen chloride | |
| Liquid |
| Colorless | |
| 5470-11-1 , 7732-18-5 | |
| ClH4NO | |
| MFCD00051089 | |
| hydroxylamine hydrochloride, hydroxylamine hcl, hydroxylammonium chloride, oxammonium hydrochloride, hydroxylamine, hydrochloride, hydroxyamine hydrochloride, hydroxylamine chloride 1:1, hydroxylamine, hydrochloride 1:1, hydroxyamine hcl, oxammonium hci | |
| WTDHULULXKLSOZ-UHFFFAOYSA-N | |
| hydroxylamine hydrochloride | |
| 443297 | |
| 69.49 | |
| Certified | |
| Poly Bottle | |
| 500 mL |
Chemical Identifiers
| 5470-11-1 | |
| 69.49 | |
| WTDHULULXKLSOZ-UHFFFAOYSA-N | |
| 443297 | |
| hydroxylamine hydrochloride |
| ClH4NO | |
| MFCD00051089 | |
| hydroxylamine hydrochloride, hydroxylamine hcl, hydroxylammonium chloride, oxammonium hydrochloride, hydroxylamine, hydrochloride, hydroxyamine hydrochloride, hydroxylamine chloride 1:1, hydroxylamine, hydrochloride 1:1, hydroxyamine hcl, oxammonium hci | |
| CHEBI:5807 | |
| Cl.NO |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Toxic in contact with skin.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
Suspected of causing cancer.
May cause damage to organs (blood) through prolonged or repeated exposure.
Very toxic to aquatic life.
GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Do not breathe mist.
Do not eat, drink or smoke when using this product.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Contaminated work clothing must not be allowed out of the workplace.
Avoid release to the environment.
If exposed or concerned: Get medical advice/attention.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
If on skin: Wash skin with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
Take off contaminated clothing and wash it before reuse.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Collect spillage.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Danger
Recommended Storage : Room Temperature