missing translation for 'onlineSavingsMsg'
Learn More

Hydroxylamine Hydrochloride, 50 g + 100 mL Water, Ricca Chemical

Supplier:  Ricca Chemical Company 39254

 View more versions of this product

Catalog No. 3925-4


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Hydroxylamine Hydrochloride
7732-18-5
Laboratory
3.2
Liquid
Colorless
dihydrogen oxide, dihydrogen monoxide
Natural Poly Bottle
120 mL
50g + 100mL Water

Chemical Identifiers

7732-18-5
dihydrogen oxide, dihydrogen monoxide
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions