missing translation for 'onlineSavingsMsg'
Learn More

Hydroxylamine Hydrochloride, 10% (w/w), Ricca Chemical

Catalog No. 390016
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
390016 500 mL Natural Poly Bottle
390032 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 390016 Supplier Ricca Chemical Company Supplier No. 390016

Chemical Identifiers

CAS 5470-11-1 , 7732-18-5
Molecular Formula ClH4NO
Molecular Weight (g/mol) 69.49
MDL Number MFCD00051089
InChI Key WTDHULULXKLSOZ-UHFFFAOYSA-N
Synonym hydroxylamine hydrochloride, hydroxylamine hcl, hydroxylammonium chloride, oxammonium hydrochloride, hydroxylamine, hydrochloride, hydroxyamine hydrochloride, hydroxylamine chloride 1:1, hydroxylamine, hydrochloride 1:1, hydroxyamine hcl, oxammonium hci
PubChem CID 443297
ChEBI CHEBI:5807
IUPAC Name hydroxylamine hydrochloride
SMILES Cl.NO

Specifications

Color Colorless
CAS Min % 88.0, 9.81
CAS Max % 92.0, 10.21
Percent Purity 10%
Grade Laboratory
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
pH 3.2
Quantity 500 mL
Physical Form Liquid
Concentration or Composition Notes 10% (w/w)
Chemical Name or Material Hydroxylamine Hydrochloride
Show More Show Less