missing translation for 'onlineSavingsMsg'
Learn More

Hydroxylamine Hydrochloride, 10% (w/v), Ricca Chemical

Supplier:  Ricca Chemical Company R39010001A

 View more versions of this product

Catalog No. R39010001A


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Hydroxylamine Hydrochloride, 10% (w/v)
5470-11-1,7732-18-5
90.48,9.52
90.48,9.52
MFCD00051089
hydroxylamine hydrochloride, hydroxylamine hcl, hydroxylammonium chloride, oxammonium hydrochloride, hydroxylamine, hydrochloride, hydroxyamine hydrochloride, hydroxylamine chloride 1:1, hydroxylamine, hydrochloride 1:1, hydroxyamine hcl, oxammonium hci
WTDHULULXKLSOZ-UHFFFAOYSA-N
hydroxylamine hydrochloride
443297
10%
Natural Poly Bottle
1 L
Colorless
∼100°C
∼0°C
ClH4NO
1.05
Miscible in water
Cl.NO
69.49
CHEBI:5807
Laboratory
3.2 (1.4% solution)
Liquid

Chemical Identifiers

5470-11-1
69.49
WTDHULULXKLSOZ-UHFFFAOYSA-N
443297
hydroxylamine hydrochloride
ClH4NO
MFCD00051089
hydroxylamine hydrochloride, hydroxylamine hcl, hydroxylammonium chloride, oxammonium hydrochloride, hydroxylamine, hydrochloride, hydroxyamine hydrochloride, hydroxylamine chloride 1:1, hydroxylamine, hydrochloride 1:1, hydroxyamine hcl, oxammonium hci
CHEBI:5807
Cl.NO
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

ShelfLife : 12 months

Recommended Storage : Ambient Corrosive

SDS
Documents

Documents

Promotions

Promotions