missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydroxylamine Hydrochloride, 10% (w/v), Ricca Chemical
$103.59 - $204.16
Chemical Identifiers
| CAS | 5470-11-1 |
|---|---|
| Molecular Formula | ClH4NO |
| Molecular Weight (g/mol) | 69.49 |
| MDL Number | MFCD00051089 |
| InChI Key | WTDHULULXKLSOZ-UHFFFAOYSA-N |
| Synonym | hydroxylamine hydrochloride, hydroxylamine hcl, hydroxylammonium chloride, oxammonium hydrochloride, hydroxylamine, hydrochloride, hydroxyamine hydrochloride, hydroxylamine chloride 1:1, hydroxylamine, hydrochloride 1:1, hydroxyamine hcl, oxammonium hci |
| PubChem CID | 443297 |
| ChEBI | CHEBI:5807 |
| IUPAC Name | hydroxylamine hydrochloride |
| SMILES | Cl.NO |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
R3901000500
|
Ricca Chemical Company
R3901000500A |
500 mL | Natural Poly Bottle |
Each for $103.59
|
|
||||
|
R39010001A
|
Ricca Chemical Company
R39010001A |
1 L | Natural Poly Bottle |
Each for $204.16
|
|
||||
Chemical Identifiers
| 5470-11-1 | |
| 69.49 | |
| WTDHULULXKLSOZ-UHFFFAOYSA-N | |
| 443297 | |
| hydroxylamine hydrochloride |
| ClH4NO | |
| MFCD00051089 | |
| hydroxylamine hydrochloride, hydroxylamine hcl, hydroxylammonium chloride, oxammonium hydrochloride, hydroxylamine, hydrochloride, hydroxyamine hydrochloride, hydroxylamine chloride 1:1, hydroxylamine, hydrochloride 1:1, hydroxyamine hcl, oxammonium hci | |
| CHEBI:5807 | |
| Cl.NO |
Specifications
| Colorless | |
| ∼100°C | |
| ∼0°C | |
| ClH4NO | |
| 1.05 | |
| Miscible in water | |
| Cl.NO | |
| 69.49 | |
| CHEBI:5807 | |
| Laboratory | |
| 3.2 (1.4% solution) | |
| Liquid |
| 5470-11-1,7732-18-5 | |
| 90.48,9.52 | |
| 90.48,9.52 | |
| MFCD00051089 | |
| hydroxylamine hydrochloride, hydroxylamine hcl, hydroxylammonium chloride, oxammonium hydrochloride, hydroxylamine, hydrochloride, hydroxyamine hydrochloride, hydroxylamine chloride 1:1, hydroxylamine, hydrochloride 1:1, hydroxyamine hcl, oxammonium hci | |
| WTDHULULXKLSOZ-UHFFFAOYSA-N | |
| hydroxylamine hydrochloride | |
| 443297 | |
| 10% | |
| Natural Poly Bottle | |
| 500 mL | |
| Hydroxylamine Hydrochloride, 10% (w/v) |
Safety and Handling
ShelfLife : 12 months
Recommended Storage : Ambient Corrosive