missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydroxy Naphthol Blue, ACS Reagent Grade, Ricca Chemical
Deposited on Sodium Chloride crystals
$159.20 - $2,720.33
Chemical Identifiers
| CAS | 165660-27-5 |
|---|---|
| Molecular Formula | C20H14N2Na2O11S3 |
| Molecular Weight (g/mol) | Mixture |
| InChI Key | LXJHFXHZWYBBKJ-LUUCHEBKSA-N |
| PubChem CID | 131849366 |
| IUPAC Name | 3-hydroxy-4-[(2Z)-2-(2-oxo-4-sulfonaphthalen-1-ylidene)hydrazinyl]naphthalene-2,7-disulfonic acid;sodium |
| SMILES | C1=CC=C2C(=C1)C(=CC(=O)C2=NNC3=C4C=CC(=CC4=CC(=C3O)S(=O)(=O)O)S(=O)(=O)O)S(=O)(=O)O.[Na].[Na] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
393025
|
N/A | 25 g | Pouch |
Each for $159.20
|
|
||||
|
3930100
|
N/A | 100 g | White Poly Wide Mouth Bottle |
Each for $389.50
|
|
||||
|
39301KG
|
N/A | 1000 gal. | White Poly Wide Mouth Bottle |
Each for $2,720.33
|
|
||||
Chemical Identifiers
| 165660-27-5 | |
| Mixture | |
| 131849366 | |
| C1=CC=C2C(=C1)C(=CC(=O)C2=NNC3=C4C=CC(=CC4=CC(=C3O)S(=O)(=O)O)S(=O)(=O)O)S(=O)(=O)O.[Na].[Na] |
| C20H14N2Na2O11S3 | |
| LXJHFXHZWYBBKJ-LUUCHEBKSA-N | |
| 3-hydroxy-4-[(2Z)-2-(2-oxo-4-sulfonaphthalen-1-ylidene)hydrazinyl]naphthalene-2,7-disulfonic acid;sodium |
Specifications
| 165660-27-5 | |
| 100.0 | |
| LXJHFXHZWYBBKJ-LUUCHEBKSA-N | |
| 3-hydroxy-4-[(2Z)-2-(2-oxo-4-sulfonaphthalen-1-ylidene)hydrazinyl]naphthalene-2,7-disulfonic acid;sodium | |
| 131849366 | |
| Dark Blue | |
| Powder |
| 98.0 | |
| C20H14N2Na2O11S3 | |
| C1=CC=C2C(=C1)C(=CC(=O)C2=NNC3=C4C=CC(=CC4=CC(=C3O)S(=O)(=O)O)S(=O)(=O)O)S(=O)(=O)O.[Na].[Na] | |
| Mixture | |
| ACS | |
| 7.9 | |
| Hydroxy Naphthol Blue, In NaCl |