missing translation for 'onlineSavingsMsg'
Learn More

Hydroxy Naphthol Blue, ACS Reagent Grade, Ricca Chemical

Catalog No. 3930100
Change view
Click to view available options
Quantity:
25 g
100 g
1000 gal.
Packaging:
Pouch
White Poly Wide Mouth Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
3930100 100 g White Poly Wide Mouth Bottle
393025 25 g Pouch
39301KG 1000 gal. White Poly Wide Mouth Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 3930100 Supplier Ricca Chemical Company Supplier No. 3930100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Deposited on Sodium Chloride crystals

Chemical Identifiers

CAS 165660-27-5
Molecular Formula C20H14N2Na2O11S3
Molecular Weight (g/mol) Mixture
InChI Key LXJHFXHZWYBBKJ-LUUCHEBKSA-N
PubChem CID 131849366
IUPAC Name 3-hydroxy-4-[(2Z)-2-(2-oxo-4-sulfonaphthalen-1-ylidene)hydrazinyl]naphthalene-2,7-disulfonic acid;sodium
SMILES C1=CC=C2C(=C1)C(=CC(=O)C2=NNC3=C4C=CC(=CC4=CC(=C3O)S(=O)(=O)O)S(=O)(=O)O)S(=O)(=O)O.[Na].[Na]

Specifications

CAS Min % 98.0
CAS Max % 100.0
Packaging White Poly Wide Mouth Bottle
Color Dark Blue
pH 7.9
Quantity 100 g
Physical Form Powder
Grade ACS
Chemical Name or Material Hydroxy Naphthol Blue, In NaCl