accessibility menu, dialog, popup

UserName

ONNHHOSOOOHSOOHOSOOHONaNa(E/Z)

CAS RN 165660-27-5

IUPAC Name: 3-hydroxy-4-[2-(2-oxo-4-sulfo-1,2-dihydronaphthalen-1-ylidene)hydrazin-1-yl]naphthalene-2,7-disulfonic acid disodium
Synonyms: 3-hydroxy-4-[2-(2-oxo-4-sulfonaphthalen-1-ylidene)hydrazin-1-yl]naphthalene-2,7-disulfonic acid disodium
Molecular Weight (g/mol): 600.49
Molecular Formula: C20H14N2Na2O11S3
InChi Key: LXJHFXHZWYBBKJ-UHFFFAOYSA-N
SMILES: [Na].[Na].OC1=C(NN=C2C(=O)C=C(C3=CC=CC=C23)S(O)(=O)=O)C2=CC=C(C=C2C=C1S(O)(=O)=O)S(O)(=O)=O
Color 
  • (1)
  • (3)
  • (2)
  • (5)
Molecular Weight (g/mol) 
  • (2)
  • (4)
  • (2)
  • (3)
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Grade 
  • (7)
  • (3)
pH 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (3)
  • (1)
  • (3)
  • (2)
  • (5)
  • (2)
  • (4)
  • (2)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (7)
  • (3)
  • (3)