missing translation for 'onlineSavingsMsg'
Learn More

HydroSpec™ Solvent C, Ricca Chemical

Formulated for high performance and reliable results. Karl Fischer reagent for moisture determination.

$466.54 - $466.54

Specifications

Chemical Name or Material Solvent C
CAS Min % 5,20
CAS Max % 10,10
CAS 7446-09-5 , 67-56-1 , 67-66-3 , 288-32-4
Physical Form Liquid
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
RK3300001C
View Documents
Ricca Chemical Company
RK3300001C
1 L
Each for $466.54
Only null left
 
Description

Description

Specifications

Specifications

Solvent C
10,10
Liquid
UN1992
Karl Fischer
5,20
7446-09-5 , 67-56-1 , 67-66-3 , 288-32-4
Amber Glass Bottle
1.04
Videos
SDS
Documents

Documents