missing translation for 'onlineSavingsMsg'
Learn More

Hydroquinone-O,O'-diacetic acid, 98%

Catalog No. AAB2264522
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2264522 100 g
AAB2264506 5 g
AAB2264514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2264522 Supplier Thermo Scientific Chemicals Supplier No. B2264522
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2245-53-6 | C10H10O6 | 226.18 g/mol

Hydroquinone-O,O'-diacetic acid is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hydroquinone-O,O′-diacetic acid is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 2245-53-6
Molecular Formula C10H10O6
Molecular Weight (g/mol) 226.18
MDL Number MFCD00016816
InChI Key DNXOCFKTVLHUMU-UHFFFAOYSA-N
Synonym 2,2'-1,4-phenylenebis oxy diacetic acid, 1,4-phenylenedioxydiacetic acid, hydroquinone-o,o'-diacetic acid, 1,4-dicarboxymethoxybenzene, hydroquinone-o,o-diacetic acid, acetic acid, p-phenylenedioxy di, acetic acid, 2,2'-1,4-phenylenebis oxy bis, 2-4-carboxymethoxy phenoxy acetic acid
PubChem CID 75251
IUPAC Name 2-[4-(carboxymethoxy)phenoxy]acetic acid
SMILES OC(=O)COC1=CC=C(OCC(O)=O)C=C1

Specifications

Melting Point ∼255°C (decomposition)
Quantity 100 g
Beilstein 1984454
Solubility Information Slightly soluble in water.
Formula Weight 226.19
Percent Purity 98%
Chemical Name or Material Hydroquinone-O,O'-diacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 218-834-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only