accessibility menu, dialog, popup

UserName

CAS RN 2245-53-6

IUPAC Name: 2-[4-(carboxymethoxy)phenoxy]acetic acid
Synonyms: 4-(carboxymethoxy)phenoxyacetic acid
Molecular Weight (g/mol): 226.18
Molecular Formula: C10H10O6
InChi Key: DNXOCFKTVLHUMU-UHFFFAOYSA-N
SMILES: OC(=O)COC1=CC=C(OCC(O)=O)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results