missing translation for 'onlineSavingsMsg'
Learn More

Hydroquinone, 1% (w/v) (10 g/L), Ricca Chemical

Catalog No. p-3251800
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
386516 500 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 386516 Supplier Ricca Chemical Company Supplier No. 386516
Only null left

Chemical Identifiers

CAS 123-31-9 , 7732-18-5
Molecular Formula C6H6O2
Molecular Weight (g/mol) 110.112
InChI Key QIGBRXMKCJKVMJ-UHFFFAOYSA-N
Synonym hydroquinone, 1,4-benzenediol, quinol, 1,4-dihydroxybenzene, p-benzenediol, 4-hydroxyphenol, p-hydroquinone, p-hydroxyphenol, p-dihydroxybenzene, benzoquinol
PubChem CID 785
ChEBI CHEBI:17594
IUPAC Name benzene-1,4-diol
SMILES C1=CC(=CC=C1O)O

Specifications

Color Colorless
CAS Min % 97.0, 0.97
CAS Max % 100.0, 1.01
Percent Purity 1%
Grade Laboratory
Packaging Amber Glass Bottle
Solvent or Matrix Aqueous Solution
Quantity 500 mL
Physical Form Liquid
Concentration or Composition Notes 1% (w/v),10g/L
Chemical Name or Material Hydroquinone
Show More Show Less