missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen Sulfide Water, Saturated, Ricca Chemical

Catalog No. 385016
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
385016 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 385016 Supplier Ricca Chemical Company Supplier No. 385016
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Meets USP and ACS RS Specifications

Chemical Identifiers

CAS 7732-18-5

Specifications

Color Colorless
CAS Min % 98.0
CAS Max % 100.0
Grade Laboratory
Packaging Amber Glass Bottle
pH 4.5
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Hydrogen Sulfide Water, Saturated