missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrogen Sulfide Water, Saturated, Ricca Chemical
Meets USP and ACS RS Specifications
$121.72 - $428.33
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
36-101-9130
|
Ricca Chemical Company
RI38504 |
120 mL |
Each for $428.33
|
|
|||||
|
3850-32
|
Ricca Chemical Company
385032 |
1 L |
Each for $121.72
|
|
|||||
Chemical Identifiers
| 7732-18-5 | |
| dihydrogen oxide, dihydrogen monoxide |
Specifications
| 7732-18-5 | |
| 100.0 | |
| Laboratory | |
| 4.5 | |
| HYDROGEN SULFIDE WATER |
| 98.0 | |
| dihydrogen oxide, dihydrogen monoxide | |
| Amber Glass Bottle | |
| 120 mL |