Learn More
Hydrogen peroxide, 35% w/w aq. soln., stab.
CAS: 7722-84-1 | H2O2 | 34.014 g/mol
$68.89 - $104.70
Chemical Identifiers
| CAS | 7722-84-1 |
|---|---|
| Molecular Formula | H2O2 |
| MDL Number | MFCD00011333 |
| InChI Key | MHAJPDPJQMAIIY-UHFFFAOYSA-N |
| Synonym | oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone |
| PubChem CID | 784 |
| ChEBI | CHEBI:16240 |
| IUPAC Name | hydrogen peroxide |
| SMILES | OO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL14000BEX
|
Thermo Scientific Chemicals
L14000BEX |
2 x 50 mL |
N/A
|
N/A | |||||
|
Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. |
|||||||||
|
AAL14000AP
|
Thermo Scientific Chemicals
L14000AP |
500 mL |
N/A
|
N/A | |||||
|
Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. |
|||||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Chemical Identifiers
| 7722-84-1 | |
| MFCD00011333 | |
| oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone | |
| CHEBI:16240 | |
| OO |
| H2O2 | |
| MHAJPDPJQMAIIY-UHFFFAOYSA-N | |
| 784 | |
| hydrogen peroxide |
Specifications
| 7722-84-1 | |
| -26°C | |
| MFCD00011333 | |
| 3587191 | |
| 1.13 | |
| oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone | |
| MHAJPDPJQMAIIY-UHFFFAOYSA-N | |
| hydrogen peroxide | |
| 784 | |
| 34.02 | |
| 1.335 | |
| Hydrogen peroxide, stabilized |
| 107°C | |
| H2O2 | |
| UN2014 | |
| 35% w/w aq. soln. | |
| 14,4798 | |
| Miscible with water,ether,alcohol,organic solvents and carboxylic esters. Immiscible with hydrocarbons and petroleum ether. | |
| OO | |
| 34.014 | |
| CHEBI:16240 | |
| Pungent | |
| 2 x 50 mL |
Safety and Handling
GHS H Statement
H272-H314-H318-H302-H335
May intensify fire; oxidizer.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
P210-P220-P221-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P362-P370+P378q-P501c
H270-H302-H314-H332-H335
DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: HYDROGEN PEROXIDE, AQUEOUS SOLUTIONS
EINECSNumber : 231-765-0
RTECSNumber : MX0899000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only