missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen peroxide, 35% w/w aq. soln., stab.

CAS: 7722-84-1 | H2O2 | 34.014 g/mol

Supplier:  Thermo Scientific Chemicals L14000BEX

 View more versions of this product

Catalog No. AAL14000BEX

Due to product restrictions, we cannot sell this product online.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Specifications

Specifications

Hydrogen peroxide
7722-84-1
-26°C
MFCD00011333
3587191
1.13
oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
MHAJPDPJQMAIIY-UHFFFAOYSA-N
hydrogen peroxide
784
34.02
1.335
stabilized
107°C
H2O2
UN2014
35% w/w aq. soln.
14,4798
Miscible with water,ether,alcohol,organic solvents and carboxylic esters. Immiscible with hydrocarbons and petroleum ether.
OO
34.014
CHEBI:16240
Pungent
2 x 50 mL

Chemical Identifiers

7722-84-1
MFCD00011333
oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
CHEBI:16240
OO
H2O2
MHAJPDPJQMAIIY-UHFFFAOYSA-N
784
hydrogen peroxide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H272-H314-H318-H302-H335
May intensify fire; oxidizer.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.

P210-P220-P221-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P362-P370+P378q-P501c

H270-H302-H314-H332-H335

DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: HYDROGEN PEROXIDE, AQUEOUS SOLUTIONS

EINECSNumber : 231-765-0

RTECSNumber : MX0899000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only