missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen Peroxide, 35% Solution, Technical, 30-36%, Spectrum™ Chemical

Catalog No. 18610955
Change view
Click to view available options
Quantity:
500 mL
4 L
200 L
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610955 500 mL Poly Bottle
18610962 4 L Poly Bottle
18610956 200 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18610955 Supplier Spectrum Chemical Mfg Cor Supplier No. H1075500MLPL

Due to product restrictions, we cannot sell this product online.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7722-84-1 , 7732-18-5

Specifications

CAS Max % 1
Percent Purity 30 to 36%
Grade Technical
Packaging Poly Bottle
Quantity 500 mL
Chemical Name or Material Hydrogen Peroxide

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.