missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen Peroxide, 35% Solution, FCC, Spectrum™ Chemical

Catalog No. 18603264
Change view
Click to view available options
Quantity:
4 L
6 x 500 mL
4 x 4 L
200 L
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603264 6 x 500 mL Poly Bottle
18603265 4 L Poly Bottle
18603266 4 x 4 L Poly Bottle
18603267 200 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18603264 Supplier Spectrum Chemical Mfg Cor Supplier No. HY115500MLPLCS6

Due to product restrictions, we cannot sell this product online.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

HY115, 7722-84-1, H2O2

Spectrum™ Chemical Hydrogen Peroxide, 35 Percent Solution, FCC is used as a disinfectant in food production. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7732-18-5 , 7722-84-1
Molecular Formula H2O2
Molecular Weight (g/mol) 34.01
InChI Key MHAJPDPJQMAIIY-UHFFFAOYSA-N
IUPAC Name peroxol
SMILES OO

Specifications

Acidity 0.03%
CAS Max % 65%, 35%
UN Number UN2014
Concentration 35%
Percent Purity 35%
Grade FCC
Packaging Poly Bottle
Quantity 6 x 500 mL
Chemical Name or Material Hydrogen Peroxide