missing translation for 'onlineSavingsMsg'
Learn More

hydrogen peroxide, 3 wt.% solution in water, stabilized

CAS: 7722-84-1 | H2O2 | 34.014 g/mol

Supplier:  Thermo Scientific Chemicals 426000010

 View more versions of this product

Catalog No. AC426000010


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
Specifications

Specifications

Hydrogen peroxide
7722-84-1 , 7732-18-5
H2O2
MFCD00011333
15, 4835
Solubility in water: soluble.
OO
34.014
CHEBI:16240
Plastic bottle
1 L
3 wt.% solution in water, stabilized
100.0°C
HOOH
1.0100g/mL
oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
MHAJPDPJQMAIIY-UHFFFAOYSA-N
hydrogen peroxide
784
34
1.01

Chemical Identifiers

7722-84-1
34.014
MHAJPDPJQMAIIY-UHFFFAOYSA-N
784
hydrogen peroxide
H2O2
MFCD00011333
oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
CHEBI:16240
OO
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only