missing translation for 'onlineSavingsMsg'
Learn More

hydrogen peroxide, 3 wt.% solution in water, stabilized

Catalog No. AC426000250
Change view
Click to view available options
Quantity:
25 mL
100 mL
1 L
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC426000250 25 mL Plastic bottle
AC426001000 100 mL Plastic bottle
AC426000010 1 L Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC426000250 Supplier Thermo Scientific Chemicals Supplier No. 426000250

CAS: 7722-84-1 | H2O2 | 34.014 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent

Chemical Identifiers

CAS 7722-84-1 , 7732-18-5
Molecular Formula H2O2
Molecular Weight (g/mol) 34.014
MDL Number MFCD00011333
InChI Key MHAJPDPJQMAIIY-UHFFFAOYSA-N
Synonym oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
PubChem CID 784
ChEBI CHEBI:16240
IUPAC Name hydrogen peroxide
SMILES OO

Specifications

Boiling Point 100.0°C
Linear Formula HOOH
Density 1.0100g/mL
Merck Index 15, 4835
Solubility Information Solubility in water: soluble.
Formula Weight 34
Packaging Plastic bottle
Specific Gravity 1.01
Quantity 25 mL
Chemical Name or Material Hydrogen peroxide, 3 wt.% solution in water, stabilized
Hazard Statement GHS H Statement:
Causes skin irritation.

RUO – Research Use Only