missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen Peroxide, 10% (w/w), Reagent Grade, Ricca Chemical

Catalog No. R38213101BV
Change view
Click to view available options
Quantity:
1 L
Packaging:
Vented Opaque Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R38213101BV 1 L Vented Opaque Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R38213101BV Supplier Ricca Chemical Company Supplier No. R38213101BV

Stabilized with Acetanilide

Chemical Identifiers

CAS 7722-84-1 , 7732-18-5
Molecular Formula H2O2
Molecular Weight (g/mol) 34.014
InChI Key MHAJPDPJQMAIIY-UHFFFAOYSA-N
Synonym oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
PubChem CID 784
ChEBI CHEBI:16240
IUPAC Name hydrogen peroxide
SMILES OO

Specifications

Color Colorless
Percent Purity 10%
Grade Reagent
Packaging Vented Opaque Poly Bottle
Quantity 1 L
Physical Form Liquid
Chemical Name or Material Hydrogen Peroxide, 10% (w/w) Aqueous Solution, Stabilized with Acetanilide