missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen Peroxide, 10% (v/v) (Approx. 3% by weight), unstabilized Technical Grade, Ricca Chemical

$616.67 - $2661.81

Chemical Identifiers

CAS 7722-84-1
Molecular Formula H2O2
Molecular Weight (g/mol) 34.014
InChI Key MHAJPDPJQMAIIY-UHFFFAOYSA-N
Synonym oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
PubChem CID 784
ChEBI CHEBI:16240
IUPAC Name hydrogen peroxide
SMILES OO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
382132
View Documents
Ricca Chemical Company
382132
1 L Natural Poly Bottle
Each for $616.67
Only null left
 
3821255
View Documents
Ricca Chemical Company
3821255
55 gal. Poly Drum
Each for $2,661.81
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7722-84-1
34.014
oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
CHEBI:16240
OO
H2O2
MHAJPDPJQMAIIY-UHFFFAOYSA-N
784
hydrogen peroxide

Specifications

Colorless
88.0,9.79
H2O2
MHAJPDPJQMAIIY-UHFFFAOYSA-N
hydrogen peroxide
784
10%
Natural Poly Bottle
1 L
10% (v/v),Approximately 3% by weight
7722-84-1,7732-18-5
92.0,10.19
oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
OO
34.014
CHEBI:16240
Technical
Aqueous Solution
Liquid
Hydrogen Peroxide
Videos
SDS
Documents

Documents