missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen chloride, 5 to 6M soln. in 2-propanol, pure

Catalog No. AAH31519AE
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH31519AE 100 mL
AAH31519AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAH31519AE Supplier Thermo Scientific Chemicals Supplier No. H31519AE
Only null left
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 7647-01-0 | ClH | 36.46 g/mol

Hydrogen chloride, 5 to 6M soln. in 2-propanol may be used in measuring their conductance at 250. It is used in the manufacturing of pharmaceuticals to get a hydrochloride salt. For example, ranitidine base is converted to ranitidine hydrochloride, which is used as anti ulcerative drug.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
SMILES Cl

Specifications

Linear Formula HCl
UN Number UN2924
Concentration 5 to 6M soln. in 2-propanol
Density 0.909
Solubility Information Miscible with water,methanol,ethanol and 2-propanol.
IUPAC Name chlorane
Formula Weight 36.46
Odor Alcohol-like
Quantity 100 mL
Flash Point 11°C (52°F)
Sensitivity Hygroscopic
Chemical Name or Material Hydrochloric acid
Grade Pure
Show More Show Less
Hazard Category H225-H314-H332-H335-H336
Hazard Statement GHS H Statement
H225-H314-H336
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.
Precautionary Statement P210-P233-P234-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P390-P501c
DOTInformation Hazard Class: 8; Packaging Group: I
EINECSNumber 231-595-7
TSCA Yes
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only