Learn More
Hydrogen chloride, 3M in cyclopentyl methyl ether
Description
In chemical industry production of inorganic salts, Production of vinyl chloride from acetylene, alkyl chlorides from olefins, dyes, artificial silk, pigment, pesticides, pharmaceutical hydrochloride, As catalyst and solvent in organic syntheses, Alcohol chlorination reagent, Nitration reaction, Manufacture of phosphoric acid and in the production of ammonium chloride, Neutralization of basic systems, Cleaning agents. Hydrolyzing of starch and proteins in the preparation of various food products. The hydrogen chloride gas is mainly used for EDC and vinyl chloride monomer (VCM) production. CPME has proved to be quite useful as a process solvent for organic synthesis, simply because it is free of the drawbacks present in the more traditional solvents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 106°C |
| Linear Formula | HCl |
| UN Number | UN2924 |
| Concentration | 3M in cyclopentyl methyl ether |
| Density | 0.895 |
| Solubility Information | Insoluble in water. |
| IUPAC Name | hydrogen chloride |
| Formula Weight | 36.46 |
| Quantity | 100 mL |
| Flash Point | 10°C (50°F) |
| Show More |
Safety and Handling
| Hazard Category | H225-H302-H314-H331-H335 |
| Hazard Statement | GHS H Statement H225-H331-H314-H302 Highly flammable liquid and vapor. Toxic if inhaled. Causes severe skin burns and eye damage. Harmful if swallowed. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c |
| DOTInformation | Hazard Class: 3 |
| EINECSNumber | 231-595-7 |
| RTECSNumber | MW4025000 |
| TSCA | Yes |
| Recommended Storage | Keep cold; Store under Nitrogen |
RUO – Research Use Only