missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen bromide, 33% w/w (45% w/v) soln. in acetic acid

CAS: 37348-16-6 | BrH | 80.91 g/mol

$297.81 - $1146.90

Chemical Identifiers

CAS 37348-16-6
Molecular Formula BrH
Molecular Weight (g/mol) 80.91
MDL Number MFCD00011323
InChI Key CPELXLSAUQHCOX-UHFFFAOYSA-N
Synonym hydrogen bromide, hydrobromic acid, bromwasserstoff, broomwaterstof, bromowodor, acido bromidrico, acide bromhydrique, anhydrous hydrobromic acid, bromowodor polish, hydrobromide
PubChem CID 260
ChEBI CHEBI:47266
IUPAC Name bromane
SMILES Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA14475AP
View Documents
Thermo Scientific Chemicals
A14475AP
500 mL
Each for $297.81
Only null left
 
AAA144750F
View Documents
Thermo Scientific Chemicals
A144750F
2500 mL
Each for $1,146.90
Only null left
 
Description

Description

Hydrobromic acid is an indispensable raw material for organic intermediates, inorganic pharmaceuticals, photosensitive materials, dyes and medicines. It is also used in the production of bromine compounds. It is used as an important agent in the industrial processes of isomerization, polymerization, hydration and dehydration and esterification reactions. It plays an essential role for the extraction of mercury from the cinnebar ore.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

37348-16-6
80.91
CPELXLSAUQHCOX-UHFFFAOYSA-N
260
bromane
BrH
MFCD00011323
hydrogen bromide, hydrobromic acid, bromwasserstoff, broomwaterstof, bromowodor, acido bromidrico, acide bromhydrique, anhydrous hydrobromic acid, bromowodor polish, hydrobromide
CHEBI:47266
Br

Specifications

37348-16-6
HBr
UN3265
33% w/w (45% w/v) soln. in acetic acid
14,4778
Miscible with water,alcohol and organic solvents.
Br
80.91
CHEBI:47266
Pungent
65°C (149°F)
BrH
MFCD00011323
3587158
1.4
hydrogen bromide, hydrobromic acid, bromwasserstoff, broomwaterstof, bromowodor, acido bromidrico, acide bromhydrique, anhydrous hydrobromic acid, bromowodor polish, hydrobromide
CPELXLSAUQHCOX-UHFFFAOYSA-N
bromane
260
80.92
500 mL
Hydrogen bromide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H335-H227
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Combustible liquid.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H226-H314-H318-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 233-113-0

RTECSNumber : MW3850000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only