Learn More
Hydrogen bromide, 33% w/w (45% w/v) soln. in acetic acid
CAS: 37348-16-6 | BrH | 80.91 g/mol
Supplier: Thermo Scientific Chemicals A14475AP
Description
Hydrobromic acid is an indispensable raw material for organic intermediates, inorganic pharmaceuticals, photosensitive materials, dyes and medicines. It is also used in the production of bromine compounds. It is used as an important agent in the industrial processes of isomerization, polymerization, hydration and dehydration and esterification reactions. It plays an essential role for the extraction of mercury from the cinnebar ore.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Hydrogen bromide | |
| BrH | |
| MFCD00011323 | |
| 3587158 | |
| 1.4 | |
| hydrogen bromide, hydrobromic acid, bromwasserstoff, broomwaterstof, bromowodor, acido bromidrico, acide bromhydrique, anhydrous hydrobromic acid, bromowodor polish, hydrobromide | |
| CPELXLSAUQHCOX-UHFFFAOYSA-N | |
| bromane | |
| 260 | |
| 80.92 | |
| 500 mL |
| 37348-16-6 | |
| HBr | |
| UN3265 | |
| 33% w/w (45% w/v) soln. in acetic acid | |
| 14,4778 | |
| Miscible with water,alcohol and organic solvents. | |
| Br | |
| 80.91 | |
| CHEBI:47266 | |
| Pungent | |
| 65°C (149°F) |
Chemical Identifiers
| 37348-16-6 | |
| 80.91 | |
| CPELXLSAUQHCOX-UHFFFAOYSA-N | |
| 260 | |
| bromane |
| BrH | |
| MFCD00011323 | |
| hydrogen bromide, hydrobromic acid, bromwasserstoff, broomwaterstof, bromowodor, acido bromidrico, acide bromhydrique, anhydrous hydrobromic acid, bromowodor polish, hydrobromide | |
| CHEBI:47266 | |
| Br |
Safety and Handling
GHS H Statement
H314-H335-H227
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Combustible liquid.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H226-H314-H318-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 233-113-0
RTECSNumber : MW3850000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only