Learn More
Thermo Scientific™ Hydrocortisone butyrate
Supplier: Thermo Scientific™ 465050050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| Hydrocortisone butyrate | |
| C25H36O6 | |
| BMCQMVFGOVHVNG-TUFAYURCSA-N | |
| (1R,3aS,3bS,9aR,9bS,10S,11aS)-10-hydroxy-1-(2-hydroxyacetyl)-9a,11a-dimethyl-7-oxo-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl butanoate | |
| 432.55 |
| 13609-67-1 | |
| 5 g | |
| CCCC(=O)O[C@@]1(CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3[C@@H](O)C[C@]12C)C(=O)CO | |
| 432.56 |
Chemical Identifiers
| 13609-67-1 | |
| 432.56 | |
| (1R,3aS,3bS,9aR,9bS,10S,11aS)-10-hydroxy-1-(2-hydroxyacetyl)-9a,11a-dimethyl-7-oxo-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl butanoate |
| C25H36O6 | |
| BMCQMVFGOVHVNG-TUFAYURCSA-N | |
| CCCC(=O)O[C@@]1(CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3[C@@H](O)C[C@]12C)C(=O)CO |
Safety and Handling
GHS H Statement Causes skin irritation.
Causes serious eye irritation.
GHS P Statement IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protection/face protection.
GHS Signal Word: Warning
EINECSNumber : 237-093-4
RUO â Research Use Only