missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Hydrocortisone

Catalog No. 409350
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
409350 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 409350 Supplier R&D Systems™ Supplier No. 4093/50

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A Tocris product, part of R&D Systems. Adrenal glucocorticoid; immunosuppressant

Hydrocortisone is an adrenal glucocorticoid released in the stress response. Increases glucose production by gluconeogenesis. Immunosuppressant and antidiuretic hormone. Used in protocols to generate retinal pigment epithelial (RPE) cells from PSCs.

Chemical Identifiers

CAS 50-23-7
Molecular Formula C21H30O5

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 362.46
Purity >98%
Molecular Weight (g/mol) 362.466
Chemical Name or Material Hydrocortisone
Recommended Storage Store at Room Temperature