missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, Ultratrace, 32-35%, Spectrum™ Chemical

Catalog No. 18610931
Change view
Click to view available options
Quantity:
500 mL
1 L
2 L
Packaging:
PTFE Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610931 500 mL PTFE Bottle
18610932 1 L PTFE Bottle
18610933 2 L PTFE Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18610931 Supplier Spectrum Chemical Mfg Cor Supplier No. H1135500MLTF

Chemical Identifiers

CAS 7647-01-0 , 7732-18-5
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

Packaging PTFE Bottle
Quantity 500 mL
Percent Purity 32 to 35%
Grade High Purity Solvent

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.