missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, Technical (Muriatic Acid, 20°Be), Reagents

Catalog No. 01336843
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336843 4 L
01336846 1 L
01336842 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01336843 Supplier Labchem Inc Supplier No. C51010004A
Only null left

Hydrochloric Acid, Technical (Muriatic Acid, 20°Be), Technical, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 7647-01-0
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Muriatic Acid, Chlorohydric Acid
IUPAC Name water
SMILES O

Specifications

pH <1
Color Colorless to Slightly Greenish-Yellow
Boiling Point 100°C
Packaging Poly Bottle
Specific Gravity 1.15
Quantity 4 L
Physical Form Liquid
Grade Technical
Chemical Name or Material Hydrochloric Acid, Technical (Muriatic Acid, 20°Be)