missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric acid solution, Volumetric, Reag. Ph. Eur., 0.1 M HCl (0.1N), Honeywell Fluka™

Catalog No. 60046568
Change view
Click to view available options
Quantity:
1 L
5 L
10 L
4 x 5 L
6 x 1 L
Packaging:
Plastic bottle
VOLPAC™ Container
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60046568 6 x 1 L Plastic bottle
6000732 1 L Plastic bottle
6000738 5 L Plastic bottle
6000739 5 L VOLPAC™ Container
6000731 10 L VOLPAC™ Container
60046567 4 x 5 L Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 60046568 Supplier Honeywell Inc Supplier No. 353356X1L
Only null left

Volumetric, Reag. Ph. Eur., 0.1 M HCl (0.1N)

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

Chemical Identifiers

CAS 7647-01-0 , 7732-18-5
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
SMILES Cl

Specifications

Boiling Point 100°C
Concentration 0.1 M HCl (0.1N)
CAS Max % 99.6000%, 0.4000%
Density 1.000 g/cm3
Packaging Plastic bottle
Quantity 6 x 1 L
Linear Formula HCl
UN Number UN1789
IUPAC Name hydrogen chloride
Formula Weight 36.46
Grade Reagent Ph. Eur.
Chemical Name or Material Hydrochloric acid solution, Volumetric, Reag. Ph. Eur., 0.1 M HCl (0.1N)
Show More Show Less
ShelfLife 1080 days from date of manufacture
DOTInformation Transport Hazard Class: 8; Packing Group:3: III; Proper Shipping Name: Hydrochloric acid
EINECSNumber 231-791-2
Recommended Storage Room Temp