missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric acid Solution, volumetric, 0.1 M HCl (0.1N), Endotoxin Free, 0.098-0.120 N basis, MilliporeSigma™ Supelco™

Catalog No. 111015461
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015461 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015461 Supplier Merck Emd Millipore Supplier No. 210450ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

MDL Number MFCD00011324
Synonym Hydrogen chloride solution

Specifications

Boiling Point 100°C
CAS 7647-01-0
Vapor Pressure 190 mmHg (25°C)
Quantity 50 mL
Linear Formula HCl
Formula Weight 36.46
Concentration or Composition (by Analyte or Components) 0.1 M HCl (0.1 N)