missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid Solution, NF, EP, 6.0 N, Spectrum™ Chemical
$212.11 - $16873.26
Chemical Identifiers
| CAS | 7647-01-0 |
|---|---|
| Molecular Formula | ClH |
| Molecular Weight (g/mol) | 36.46 |
| MDL Number | MFCD00011324 MFCD00792839 |
| InChI Key | VEXZGXHMUGYJMC-UHFFFAOYSA-N |
| IUPAC Name | hydrogen chloride |
| SMILES | Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
18-614-912
|
Spectrum Chemical Mfg Cor
H98031LT |
1 L |
Each for $212.11
|
|
|||||
|
18-614-913
|
Spectrum Chemical Mfg Cor
H98034LT |
4 L |
Each for $776.97
|
|
|||||
|
18-614-914
|
Spectrum Chemical Mfg Cor
H980320LT |
20 L |
Each for $2,687.71
|
|
|||||
|
18-614-915
|
Spectrum Chemical Mfg Cor
H9803200LT |
200 L |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||