missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hydrochloric Acid Solution, NF, EP, 6.0 N, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor H980320LT

 View more versions of this product

Catalog No. 18-614-914



Description

Description

Specifications

Specifications

7647-01-0
20 L
MFCD00011324 MFCD00792839
Cl
36.46
5.95 to 6.05 N
ClH
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrogen chloride
EP/NF

Chemical Identifiers

7647-01-0
36.46
VEXZGXHMUGYJMC-UHFFFAOYSA-N
Cl
ClH
MFCD00011324 MFCD00792839
hydrogen chloride
SDS
Documents

Documents

Promotions

Promotions