missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid Solution (0.2N (N/5)/Certified), Fisher Chemical™

Catalog No. p-20186
Change view
Click to view available options
Quantity:
500 mL
20 L
Packaging:
Poly Bottle
PolyPac™
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SA52500 500 mL Poly Bottle
SA5220 20 L PolyPac™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SA52500 Supplier Thermo Fisher Scientific Supplier No. FLSA52500
Only null left

Standardized at 25°C to pH 8.2 against sodium hydroxide that has been standardized against NIST™ potassium acid phthalate

Chemical Identifiers

CAS 7647-01-0 , 7732-18-5
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

pH 0.10
Concentration 0.1990 to 0.2010N
Identification Pass Test
Packaging Poly Bottle
Traceability to NIST LOT 84L Potassium acid phthalate
Quantity 500 mL
Physical Form Liquid
UN Number 1789
Grade Certified
Concentration or Composition (by Analyte or Components) 0.2N (N/5)
Chemical Name or Material Hydrochloric Acid Solution, 0.2N (N/5)
Show More Show Less
Hazard Category Signal Word: CAUTION!
Hazard Statement CAUTION!
Precautionary Statement Emergency Overview
May cause eye, skin, and respiratory tract irritation . The toxicological properties have not been fully investigated.
NFPA
Health:1 Flammability:0 Instability:0
DOTInformation DOT Class 8, : Corrosive