missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, Reagent, ACS, 36.5-38%, Spectrum™ Chemical

H1035, 7647-01-0, HCl

Supplier:  Spectrum Chemical Mfg Cor H1035500MLGL

 View more versions of this product

Catalog No. 18603204


Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Hydrochloric Acid, Reagent, ACS , is a colorless and clear, highly corrosive, highly pungent solution of hydrogen chloride in water. As an monoprotic acid and one of the six common strong mineral acids, it is the least likely to be subject to an oxidation-reduction reaction. The fact that it is available as a pure reagent make it an excellent acidifying reagent. Hydrochloric acid is also frequently used during chemical analysis to prepare samples for analysis. It reacts with basic compounds, turning them into dissolved chlorides that can then undergo analysis. As an ACS grade Reagent, Spectrum Chemical manufactured Hydrochloric Acid is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness.
Specifications

Specifications

7647-01-0
1 ppm
5 ppm
ClH
UN1789
Cl
36.46
ACS
100%
Amber Glass Bottle
500 mL
MFCD00011324 MFCD00792839
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrogen chloride
36.5 to 38%

Chemical Identifiers

7647-01-0
36.46
hydrogen chloride
ClH
VEXZGXHMUGYJMC-UHFFFAOYSA-N
Cl
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions