missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, Reagent, ACS, 36.5-38%, Spectrum™ Chemical

Catalog No. p-7157537
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
4 L
6 x 500 mL
5 gal.
4 x 4 L
6 x 2.5 L
200 L
Packaging:
Amber Glass Bottle
Drum
Glass Bottle
Poly Bottle
Poly Drum
Poly-Coated Glass Bottle
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603201 100 mL Amber Glass Bottle
18603204 500 mL Amber Glass Bottle
18603207 2.5 L Glass Bottle
18603202 2.5 L Poly-Coated Glass Bottle
18603203 6 x 500 mL Amber Glass Bottle
18603205 4 L Poly Bottle
18603209 6 x 2.5 L Glass Bottle
18603210 6 x 2.5 L Poly-Coated Glass Bottle
18603206 4 x 4 L Poly Bottle
18603211 5 gal. Poly Drum
18603208 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 18603201 Supplier Spectrum Chemical Mfg Cor Supplier No. H1035100ML
Only null left

H1035, 7647-01-0, HCl

Spectrum™ Chemical Hydrochloric Acid, Reagent, ACS , is a colorless and clear, highly corrosive, highly pungent solution of hydrogen chloride in water. As an monoprotic acid and one of the six common strong mineral acids, it is the least likely to be subject to an oxidation-reduction reaction. The fact that it is available as a pure reagent make it an excellent acidifying reagent. Hydrochloric acid is also frequently used during chemical analysis to prepare samples for analysis. It reacts with basic compounds, turning them into dissolved chlorides that can then undergo analysis. As an ACS grade Reagent, Spectrum Chemical manufactured Hydrochloric Acid is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness.

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

CAS Max % 100%
Heavy Metals (as Pb) 1 ppm
Packaging Amber Glass Bottle
Residue after Ignition 5 ppm
Quantity 100 mL
MDL Number MFCD00011324 MFCD00792839
UN Number UN1789
Percent Purity 36.5 to 38%
Grade ACS